siRNA Details
Gene Symbol: Gimap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031247.3 | NP_112537.2 | 5 | 834 | ||
GenBank mRNA | AB164418.1 | - | 5 | 834 | ||
AF337052.1 | - | 5 | 834 | |||
BC094914.1 | - | 5 | 814 | 30074259 |
Gene Symbol: Gimap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175035.5 | NP_778200.1 | 3 | 472 | ||
GenBank mRNA | AB126961.1 | - | 3 | 472 | ||
AK052694.1 | - | 3 | 405 | |||
BC095995.1 | - | 3 | 384 | 30470329 | ||
BU694320.1 | - | 3 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
