siRNA Details
Gene Symbol: Gm29719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049966.1 | - | 488 |
Gene Symbol: Sytl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031395.2 | NP_113572.1 | 1, 2 | 388 | ||
GenBank mRNA | AB057758.1 | - | 1, 2 | 388 | ||
AB057764.1 | - | 1, 2 | 388 | |||
AB057765.1 | - | 1, 2 | 388 | |||
AK079850.1 | - | 2, 3 | 735 | |||
AK131174.1 | - | 2, 3 | 722 | |||
AK156673.1 | - | 2, 3 | 712 | |||
BC022608.1 | - | 11 | 4166796 | |||
BC118936.2 | - | 1, 2 | 213 | 40091060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
