siRNA Details
Gene Symbol: Phip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081216.1 | NP_001074685.1 | 4, 5 | 379 | ||
GenBank mRNA | AB049460.1 | - | 4, 5 | 298 | ||
AJ303103.1 | - | 4, 5 | 275 | |||
AK032797.1 | - | 4, 5 | 409 | |||
AK044935.1 | - | 4, 5 | 306 | |||
AK049736.1 | - | 4, 5 | 314 | |||
AK050025.1 | - | 4, 5 | 324 | |||
AK136530.1 | - | 4, 5 | 317 | |||
BC022102.1 | - | 4, 5 | 320 | |||
BC037036.1 | - | 4, 5 | 308 | |||
BC039765.1 | - | 4, 5 | 331 | |||
BC049950.1 | - | 4, 5 | 309 |

More Information
Other Species Targeted

Gene Ontology Categories:
