siRNA Details
Gene Symbol: Nlrp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031389.2 | NP_113566.2 | 3 | 1033 | ||
GenBank mRNA | AF285581.1 | - | 1 | 301 | ||
AK133284.1 | - | 3 | 1035 | |||
AK166817.1 | - | 3 | 1014 | |||
AK166869.1 | - | 3 | 1259 | |||
AY596194.1 | - | 3 | 1033 | |||
BC120833.1 | - | 1 | 294 | 40129756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
