siRNA Details
Gene Symbol: Sytl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031395.2 | NP_113572.1 | 2 | 452 | ||
GenBank mRNA | AB057758.1 | - | 2 | 452 | ||
AB057764.1 | - | 2 | 452 | |||
AB057765.1 | - | 2 | 452 | |||
AK079850.1 | - | 3 | 799 | |||
AK131174.1 | - | 3 | 786 | |||
AK156673.1 | - | 3 | 776 | |||
BC022608.1 | - | 1 | 75 | 4166796 | ||
BC118936.2 | - | 2 | 277 | 40091060 |
Gene Symbol: Gm29719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049966.1 | - | 552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
