siRNA Details
Gene Symbol: Phip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081216.1 | NP_001074685.1 | 4, 5 | 389 | ||
GenBank mRNA | AB049460.1 | - | 4, 5 | 308 | ||
AJ303103.1 | - | 4, 5 | 285 | |||
AK032797.1 | - | 4, 5 | 419 | |||
AK044935.1 | - | 4, 5 | 316 | |||
AK049736.1 | - | 4, 5 | 324 | |||
AK050025.1 | - | 4, 5 | 334 | |||
AK136530.1 | - | 4, 5 | 327 | |||
BC022102.1 | - | 4, 5 | 330 | |||
BC037036.1 | - | 4, 5 | 318 | |||
BC039765.1 | - | 4, 5 | 341 | |||
BC049950.1 | - | 4, 5 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
