siRNA Details
Gene Symbol: Gsdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031378.3 | NP_113555.1 | 12 | 2208 | ||
GenBank mRNA | AB042406.1 | - | 11 | 2104 | ||
AK137662.1 | - | 12 | 2209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
