siRNA Details
Gene Symbol: Nlrp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031389.2 | NP_113566.2 | 3 | 1575 | ||
GenBank mRNA | AF285581.1 | - | 1 | 843 | ||
AK133284.1 | - | 3 | 1577 | |||
AK166817.1 | - | 3 | 1556 | |||
AK166869.1 | - | 3 | 1801 | |||
AY596194.1 | - | 3 | 1575 | |||
BC120833.1 | - | 1 | 836 | 40129756 |
Gene Symbol: Nlrp4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004194.2 | NP_001004194.2 | 2 | 1538 | ||
GenBank mRNA | AY596197.1 | - | 2 | 1494 | ||
BC139319.1 | - | 2 | 1538 | 8862341 | ||
BC139320.1 | - | 2 | 1538 | 8862342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
