siRNA Details
Gene Symbol: Sytl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031395.2 | NP_113572.1 | 6 | 822 | ||
NM_183370.2 | NP_899226.2 | 4 | 236 | |||
GenBank mRNA | AB057758.1 | - | 6 | 822 | ||
AB057764.1 | - | 7 | 966 | |||
AK131174.1 | - | 7 | 1156 | |||
BC022608.1 | - | 5 | 445 | 4166796 | ||
BC106102.1 | - | 4 | 236 | 3980415 | ||
BC118936.2 | - | 6 | 647 | 40091060 |
Gene Symbol: Gm29719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049966.1 | - | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
