siRNA Details
Gene Symbol: Cep41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278891.1 | - | 2 | 116 | ||
AK019270.1 | - | 2 | 105 | |||
AK132495.1 | - | 2 | 105 | |||
AK161461.1 | - | 2 | 90 | |||
AK166974.1 | - | 2 | 97 | |||
AK170213.1 | - | 2 | 129 | |||
BC031892.1 | - | 2 | 86 | 5365552 | ||
BY712226.1 | - | 2 | 105 | |||
BY736279.1 | - | 2 | 97 |

More Information
Other Species Targeted

