siRNA Details
Gene Symbol: Ferd3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033522.2 | NP_277057.1 | 1 | 411 | ||
GenBank mRNA | AF517121.1 | - | 1 | 411 | ||
BC111576.1 | - | 1 | 403 | 40039965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
