siRNA Details
Gene Symbol: TESK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007170.2 | NP_009101.2 | 2 | 481 | ||
XM_005270355.3 | XP_005270412.1 | 2 | 453 | |||
XM_006710287.2 | XP_006710350.1 | 3 | 521 | |||
XM_011540497.1 | XP_011538799.1 | 4 | 554 | |||
XM_011540500.2 | XP_011538802.1 | 2 | 453 | |||
XM_017000047.1 | XP_016855536.1 | 3 | 686 | |||
XM_017000048.1 | XP_016855537.1 | 2 | 655 | |||
XM_017000049.1 | XP_016855538.1 | 2 | 655 | |||
GenBank mRNA | AB057597.1 | - | 2 | 473 | ||
AJ132545.1 | - | 2 | 473 | |||
AK027573.1 | - | 2 | 455 | |||
AK299269.1 | - | 2 | 476 | |||
AK302494.1 | - | 2 | 513 | |||
BC022194.1 | - | 2 | 461 | |||
BC033085.1 | - | 1 | 92 | 5425494 | ||
DA990759.1 | - | 3 | 521 | |||
DB161528.1 | - | 4 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
