siRNA Details
Gene Symbol: DNAAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256714.1 | NP_001243643.1 | 12 | 1951 | ||
NM_001256715.1 | NP_001243644.1 | 12 | 1880 | |||
NM_001256716.1 | NP_001243645.1 | 12 | 1967 | |||
NM_178837.4 | NP_849159.2 | 12 | 1891 | |||
GenBank mRNA | AK093458.1 | - | 12 | 1879 | ||
AK093473.1 | - | 7 | 1440 | |||
AK097388.1 | - | 12 | 1802 | |||
BC016843.1 | - | 12 | 1710 | 3904241 | ||
BC063449.1 | - | 12 | 1842 | 5170787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
