siRNA Details
Gene Symbol: Tssk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032004.1 | NP_114393.1 | 1 | 200 | ||
GenBank mRNA | AF329484.1 | - | 1 | 200 | ||
BC049542.1 | - | 1 | 231 |

More Information
Other Species Targeted

Gene Ontology Categories:
