siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177616.1 | NP_001171087.1 | 5, 6 | 645 | ||
NM_001177617.1 | NP_001171088.1 | 5, 6 | 812 | |||
NM_001177618.1 | NP_001171089.1 | 4, 5 | 343 | |||
NM_001177619.1 | NP_001171090.1 | 5, 6 | 1010 | |||
NM_001177623.1 | NP_001171094.1 | 5, 6 | 574 | |||
NM_033264.2 | NP_150289.1 | 4, 5 | 478 | |||
GenBank mRNA | AF324451.1 | - | 4, 5 | 438 | ||
AK029467.1 | - | 5, 6 | 1011 | |||
AK032417.1 | - | 5, 6 | 571 | |||
AK032543.1 | - | 3, 4 | 258 | |||
AK034376.1 | - | 3, 4 | 191 | |||
AK036360.1 | - | 1 | 1308 | |||
AK045507.1 | - | 5, 6 | 1012 | |||
AK050613.1 | - | 5, 6 | 575 | |||
AK137974.1 | - | 5, 6 | 813 | |||
AK153892.1 | - | 6, 7 | 1293 | |||
BC053001.1 | - | 3, 4 | 309 | 5686446 | ||
BC098368.1 | - | 5, 6 | 592 | |||
BC128376.1 | - | 4, 5 | 359 |

More Information
Other Species Targeted

Gene Ontology Categories:
