siRNA Details
Gene Symbol: Npnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287101.1 | NP_001274030.1 | 3 | 459 | ||
NM_033525.3 | NP_277060.2 | 3 | 459 | |||
GenBank mRNA | AF397008.1 | - | 3 | 211 | ||
AI527763.1 | - | 3 | 339 | |||
AY035899.1 | - | 3 | 334 | |||
CK387038.1 | - | 3 | 333 |

More Information
Other Species Targeted

Gene Ontology Categories:
