siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177616.1 | NP_001171087.1 | 8 | 875 | ||
NM_001177617.1 | NP_001171088.1 | 8 | 1042 | |||
NM_001177618.1 | NP_001171089.1 | 7 | 573 | |||
NM_001177619.1 | NP_001171090.1 | 8 | 1240 | |||
NM_001177623.1 | NP_001171094.1 | 8 | 804 | |||
NM_033264.2 | NP_150289.1 | 7 | 708 | |||
GenBank mRNA | AF324451.1 | - | 7 | 668 | ||
AK029467.1 | - | 8 | 1241 | |||
AK032417.1 | - | 8 | 801 | |||
AK032543.1 | - | 6 | 488 | |||
AK034376.1 | - | 6 | 421 | |||
AK045507.1 | - | 8 | 1242 | |||
AK050613.1 | - | 8 | 805 | |||
AK137974.1 | - | 8 | 1043 | |||
AK153892.1 | - | 9 | 1523 | |||
BC053001.1 | - | 6 | 539 | 5686446 | ||
BC098368.1 | - | 8 | 822 | |||
BC128376.1 | - | 7 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
