siRNA Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185060.1 | NP_001171989.1 | 5 | 2476 | ||
NM_001185061.1 | NP_001171990.1 | 4 | 2651 | |||
NM_001185062.1 | NP_001171991.1 | 4 | 2423 | |||
NM_198098.2 | NP_932766.1 | 4 | 2796 | |||
GenBank mRNA | U41517.1 | - | 5 | 1646 | ||
U41518.1 | - | 4 | 2333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
