siRNA Details
Gene Symbol: Vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033509.3 | NP_277044.1 | 5, 6 | 1407 | ||
GenBank mRNA | AF365875.1 | - | 5, 6 | 1171 | ||
AK033128.1 | - | 6, 7 | 1447 | |||
AK046036.1 | - | 5, 6 | 1424 | |||
AK049613.1 | - | 1422 | ||||
AY035370.3 | - | 5, 6 | 1395 | |||
BC075636.1 | - | 2264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
