siRNA Details
Gene Symbol: Npnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287101.1 | NP_001274030.1 | 5, 6 | 663 | ||
NM_001287102.1 | NP_001274031.1 | 4, 5 | 612 | |||
GenBank mRNA | AF397007.1 | - | 4, 5 | 364 | ||
AF397008.1 | - | 5, 6 | 415 |

More Information
Other Species Targeted

Gene Ontology Categories:
