siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 3 | 582 | ||
GenBank mRNA | AF330197.1 | - | 3 | 578 | ||
AK036351.1 | - | 2 | 317 | |||
AK048110.1 | - | 2 | 348 | |||
AK169999.1 | - | 2 | 692 | |||
AK171932.1 | - | 2 | 519 | |||
BC069835.1 | - | 3 | 438 | 30094289 |
