siRNA Details
Gene Symbol: Gm5124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032684.1 | - | 1 | 171 | ||
BC137827.1 | - | 1 | 151 | 8860846 | ||
BM943806.1 | - | 1 | 85 |
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039351.2 | NP_001034440.1 | 1 | 278 | ||
NM_001039352.2 | NP_001034441.1 | 1 | 278 | |||
NM_001039353.2 | NP_001034442.1 | 1 | 278 | |||
NM_053086.3 | NP_444316.2 | 1 | 278 | |||
GenBank mRNA | AK014336.1 | - | 1 | 183 | ||
AK029150.1 | - | 1 | 129 | |||
AK034817.1 | - | 1 | 157 | |||
AK129040.1 | - | 1 | 93 | |||
AK147118.1 | - | 1 | 151 | |||
AK150332.1 | - | 1 | 115 | |||
AK151031.1 | - | 1 | 115 | |||
AK152941.1 | - | 1 | 115 | |||
AK166753.1 | - | 1 | 184 | |||
BC003244.1 | - | 1 | 124 | |||
BY031361.1 | - | 1 | 279 | |||
CF750495.1 | - | 1 | 110 |

More Information
Other Species Targeted

Gene Ontology Categories:
