siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 3 | 738 | ||
GenBank mRNA | AF330197.1 | - | 3 | 734 | ||
AK036351.1 | - | 2 | 473 | |||
AK048110.1 | - | 2 | 504 | |||
AK137148.1 | - | 1 | 76 | |||
AK169999.1 | - | 2 | 848 | |||
AK171932.1 | - | 2 | 675 | |||
BC069835.1 | - | 3 | 594 | 30094289 |
