siRNA Details
Gene Symbol: IL4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000418.3 | NP_000409.1 | 11 | 3368 | ||
NM_001257406.1 | NP_001244335.1 | 10 | 3235 | |||
NM_001257407.1 | NP_001244336.1 | 11 | 3479 | |||
NM_001257997.1 | NP_001244926.1 | 10 | 3230 | |||
XM_005255308.2 | XP_005255365.1 | 3 | 2306 | |||
XM_006721043.1 | XP_006721106.1 | 2 | 2405 | |||
XM_011545825.1 | XP_011544127.1 | 10 | 3273 | |||
XM_011545826.2 | XP_011544128.1 | 11 | 3324 | |||
XM_011545827.2 | XP_011544129.1 | 11 | 4563 | |||
XM_011545828.2 | XP_011544130.1 | 9 | 3041 | |||
XM_011545829.1 | XP_011544131.1 | 10 | 3202 | |||
XM_011545830.2 | XP_011544132.1 | 10 | 4412 | |||
XM_011545831.1 | XP_011544133.1 | 9 | 3121 | |||
XM_011545832.1 | XP_011544134.1 | 9 | 3054 | |||
XM_011545833.2 | XP_011544135.1 | 8 | 2972 | |||
XM_011545834.2 | XP_011544136.1 | 8 | 2891 | |||
GenBank mRNA | X52425.1 | - | 11 | 3280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
