siRNA Details
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052994.2 | NP_443720.1 | 11 | 1401 | ||
GenBank mRNA | AJ278999.1 | - | 11 | 1403 | ||
AK122233.1 | - | 11 | 1063 | |||
AK140483.1 | - | 11 | 1308 | |||
BC057324.1 | - | 11 | 1401 | 6821449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
