siRNA Details
Gene Symbol: Tspan4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252588.1 | NP_001239517.1 | 8 | 1063 | ||
NM_053082.3 | NP_444312.1 | 8 | 1226 | |||
GenBank mRNA | AK002709.1 | - | 8 | 985 | ||
AK156959.1 | - | 8 | 1182 | |||
AK172481.1 | - | 8 | 1044 | |||
AK202774.1 | - | 1 | 146 | |||
AK206458.1 | - | 1 | 169 | |||
AK209272.1 | - | 1 | 169 | |||
AK218508.1 | - | 1 | 169 | |||
BC003482.1 | - | 8 | 1006 | 2811065 | ||
BC083070.1 | - | 8 | 1222 | 6314296 |
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BE953974.1 | - | 1 | 64 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
