siRNA Details
Gene Symbol: Rbfox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110827.2 | NP_001104297.1 | 14 | 2290 | ||
NM_001110828.2 | NP_001104298.1 | 13 | 1844 | |||
NM_001110829.2 | NP_001104299.1 | 13 | 1835 | |||
NM_001110830.2 | NP_001104300.1 | 13 | 1832 | |||
NM_001286418.1 | NP_001273347.1 | 10 | 1550 | |||
NM_001286419.1 | NP_001273348.1 | 12 | 1739 | |||
NM_053104.6 | NP_444334.3 | 14 | 2302 | |||
NM_175387.3 | NP_780596.1 | 13 | 2250 | |||
GenBank mRNA | AK077594.1 | - | 14 | 2178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
