siRNA Details
Gene Symbol: Clec2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053165.5 | NP_444395.2 | 5 | 594 | ||
GenBank mRNA | AF350410.1 | - | 5 | 573 | ||
AK090364.1 | - | 5 | 594 | |||
BC021766.1 | - | 5 | 570 | 4238060 | ||
HQ713437.1 | - | 5 | 501 | |||
HQ875061.1 | - | 5 | 537 |
