siRNA Details
Gene Symbol: Pdxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039533.2 | NP_001034622.1 | 3 | 323 | ||
NM_001291017.1 | NP_001277946.1 | 3 | 323 | |||
NM_053181.3 | NP_444411.2 | 3 | 323 | |||
GenBank mRNA | AK004611.1 | - | 3 | 200 | ||
AK030613.1 | - | 3 | 154 | |||
AK033107.1 | - | 3 | 154 | |||
AK054329.1 | - | 3 | 306 | |||
AK163696.1 | - | 3 | 194 | |||
AK165587.1 | - | 3 | 158 | |||
AK170661.1 | - | 3 | 310 | |||
BC005541.1 | - | 3 | 141 | 3590316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
