siRNA Details
Gene Symbol: Magee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053201.3 | NP_444431.3 | 1 | 1453 | ||
GenBank mRNA | AF319978.1 | - | 1 | 656 | ||
AK030032.1 | - | 1 | 1447 | |||
AK051625.1 | - | 1 | 1427 | |||
AK081378.1 | - | 1 | 1429 | |||
BC059039.1 | - | 1 | 1453 | 6812964 |
