siRNA Details
Gene Symbol: Vmn1r50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053225.1 | NP_444455.1 | 1 | 803 | ||
GenBank mRNA | BC107183.2 | - | 1 | 803 | 40050746 | |
BC115706.1 | - | 1 | 802 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053225.1 | NP_444455.1 | 1 | 803 | ||
GenBank mRNA | BC107183.2 | - | 1 | 803 | 40050746 | |
BC115706.1 | - | 1 | 802 |