siRNA Details
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053243.2 | NP_444473.1 | 3 | 279 | ||
GenBank mRNA | BC094921.1 | - | 3 | 268 | 6433981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
