siRNA Details
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF305087.1 | - | 12 | 4691 | ||
AF305088.1 | - | 12 | 2165 | |||
AF305089.1 | - | 11 | 1820 | |||
AF305090.1 | - | 12 | 1856 | |||
AK089917.1 | - | 11 | 1828 | |||
AK169974.1 | - | 12 | 2416 | |||
BC004660.1 | - | 12 | 2161 | 3497913 |
