siRNA Details
Gene Symbol: AMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000481.3 | NP_000472.2 | 2 | 340 | ||
NM_001164710.1 | NP_001158182.1 | 2 | 340 | |||
NM_001164712.1 | NP_001158184.1 | 2 | 340 | |||
NR_028435.1 | 1, 2 | 326 | ||||
GenBank mRNA | AK091738.1 | - | 2 | 129 | ||
AK096062.1 | - | 1, 2 | 177 | |||
AK290600.1 | - | 2 | 120 | |||
AK293594.1 | - | 1 | 202 | |||
AK296177.1 | - | 2 | 131 | |||
AK301072.1 | - | 1, 2 | 106 | |||
BC007546.1 | - | 2 | 104 | |||
D13811.1 | - | 2 | 241 | |||
DC387800.1 | - | 2 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
