siRNA Details
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160420.2 | NP_001153892.1 | 3 | 351 | ||
NM_001160421.2 | NP_001153893.1 | 2 | 462 | |||
NM_001160422.1 | NP_001153894.1 | 2 | 462 | |||
NM_054076.3 | NP_473417.2 | 2 | 462 | |||
GenBank mRNA | AA223016.1 | - | 2 | 462 | ||
AF333980.1 | - | 2 | 461 | |||
AK053318.1 | - | 2 | 245 | |||
AK153750.1 | - | 3 | 352 | |||
AY077682.1 | - | 2 | 454 | |||
BC099435.1 | - | 2 | 295 | 30788813 | ||
BC115928.1 | - | 2 | 415 | 40104738 |
