siRNA Details
Gene Symbol: Lmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA050915.1 | - | 2 | 284 | ||
AK133586.1 | - | 2 | 173 | |||
AW494084.1 | - | 2 | 367 | |||
BC053074.1 | - | 2 | 423 | 5693266 | ||
DV662082.1 | - | 2 | 634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
