siRNA Details
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080288.2 | NP_525027.1 | 3, 4 | 451 | ||
GenBank mRNA | AF398883.1 | - | 2, 3 | 115 | ||
AK028389.1 | - | 3, 4 | 526 | |||
AK155704.1 | - | 3, 4 | 363 | |||
AK220253.1 | - | 3, 4 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
