siRNA Details
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 4 | 1243 | ||
GenBank mRNA | AF338472.1 | - | 3 | 401 | ||
AF496547.1 | - | 4 | 473 | |||
AK045508.1 | - | 4 | 1244 | |||
AK051262.1 | - | 4 | 1244 | |||
AK122276.1 | - | 4 | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
