siRNA Details
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053273.2 | NP_444503.2 | 4 | 525 | ||
GenBank mRNA | AF329682.1 | - | 4 | 468 | ||
AK152172.1 | - | 4 | 477 | |||
AK154835.1 | - | 4 | 533 | |||
AK155166.1 | - | 4 | 526 | |||
AK156393.1 | - | 4 | 478 | |||
AK168408.1 | - | 4 | 478 | |||
BC060740.1 | - | 4 | 468 | 4158813 | ||
BY740853.1 | - | 4 | 505 |
