siRNA Details
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080288.2 | NP_525027.1 | 6 | 700 | ||
GenBank mRNA | AF398883.1 | - | 5 | 364 | ||
AK028389.1 | - | 6 | 775 | |||
AK155704.1 | - | 6 | 613 | |||
AK220253.1 | - | 6 | 588 |

More Information
Other Species Targeted

Gene Ontology Categories:
