siRNA Details
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 5 | 1339 | ||
GenBank mRNA | AF338472.1 | - | 4 | 497 | ||
AF496547.1 | - | 5 | 569 | |||
AK045508.1 | - | 5 | 1340 | |||
AK051262.1 | - | 5 | 1340 | |||
AK122276.1 | - | 5 | 987 |

More Information
Other Species Targeted

Gene Ontology Categories:
