siRNA Details
Gene Symbol: Psg26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029893.1 | NP_001025064.1 | 5 | 1572 | ||
GenBank mRNA | AK131660.1 | - | 5 | 1601 | ||
BC099404.1 | - | 5 | 1572 | 30787301 |
Gene Symbol: Psg22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004152.2 | NP_001004152.1 | 5 | 1601 | ||
GenBank mRNA | BC050099.1 | - | 5 | 1501 | 30014305 | |
BC080694.1 | - | 5 | 1613 | 30433272 |
Gene Symbol: Psg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054063.4 | NP_473404.3 | 5 | 1595 | ||
GenBank mRNA | AF113598.1 | - | 5 | 1445 | ||
BC099402.1 | - | 5 | 1581 | 30787049 | ||
BC099403.1 | - | 5 | 1466 | 30787292 | ||
BC141038.1 | - | 5 | 1548 | 9055650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
