siRNA Details
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080288.2 | NP_525027.1 | 13 | 1308 | ||
GenBank mRNA | AF398883.1 | - | 12 | 972 | ||
AK028389.1 | - | 13 | 1383 | |||
AK220253.1 | - | 13 | 1196 | |||
BC031782.1 | - | 2 | 46 | 3990906 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108415, XM_008771735, HQ438489.
Gene Ontology Categories:
