siRNA Details
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080443.2 | NP_536691.2 | 6 | 1638 | ||
GenBank mRNA | AF398968.1 | - | 3 | 923 | ||
BC062948.1 | - | 4 | 956 | 30532932 | ||
BC066824.1 | - | 6 | 1229 | 30087227 | ||
BC085155.1 | - | 6 | 1673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
