siRNA Details
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 15 | 2560 | ||
GenBank mRNA | AF338472.1 | - | 14 | 1718 | ||
AF496547.1 | - | 15 | 1790 | |||
AK122276.1 | - | 15 | 2136 | |||
CK032058.1 | - | 6 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
