siRNA Details
Gene Symbol: Ftcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080845.2 | NP_543121.1 | 3, 4 | 417 | ||
GenBank mRNA | BB610765.1 | - | 418 | |||
BC010813.1 | - | 3, 4 | 379 | 4211065 | ||
BC024078.1 | - | 3, 4 | 411 | 5098529 | ||
CD297973.1 | - | 3, 4 | 420 |
