siRNA Details
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF345951.1 | - | 2 | 745 | ||
AF345952.1 | - | 2 | 745 | |||
AF345953.1 | - | 2 | 745 | |||
AF345954.1 | - | 2 | 745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
