siRNA Details
Gene Symbol: PNKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007254.3 | NP_009185.2 | 14 | 1364 | ||
GenBank mRNA | AF120499.1 | - | 14 | 1353 | ||
AF125807.1 | - | 13 | 1254 | |||
AF126486.1 | - | 13 | 1254 | |||
BC002519.2 | - | 13 | 1212 | 3140712 | ||
BC033822.1 | - | 14 | 1360 | 5218400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
