siRNA Details
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080558.4 | NP_542125.3 | 16 | 3699 | ||
GenBank mRNA | AB093303.1 | - | 16 | 3574 | ||
AB120565.1 | - | 16 | 3584 | |||
AK037213.1 | - | 14 | 3409 | |||
AK169543.1 | - | 17 | 3877 | |||
BC008560.1 | - | 9 | 2672 | |||
BC018526.1 | - | 6 | 903 | |||
BC079535.1 | - | 10 | 3078 | |||
BC115799.1 | - | 3 | 485 | |||
BC122519.1 | - | 16 | 3585 | |||
BC141883.1 | - | 16 | 3584 | 40097831 |
