siRNA Details
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130879.2 | NP_570949.2 | 2 | 350 | ||
GenBank mRNA | AK052297.1 | - | 2 | 291 | ||
AK133251.1 | - | 3 | 367 | |||
AK166158.1 | - | 2 | 353 | |||
AK220190.1 | - | 2 | 311 | |||
BC021769.1 | - | 2 | 309 | |||
BC145295.1 | - | 3 | 341 | |||
BY734570.1 | - | 2 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
